WELL PLATE HOLDER OPTIMISED FOR INTERGRATION OF THE CERTUS FLEX INTO AN AUTOMATED LABORATORY ENVIRONMENT ALLOWS IMPROVED INSERTION A ND REMOVAL OF WELL PLATES BY A ROBOT CHAMFERED CORNER GUIDES MAXIMUM WELL PLATE HIGH 44 MM 1 * 1 items
Catalog # MSPP21192
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together