BioCircuit MEA 48 plate,16 microelectrodes per well including one large stimulation electrode per well, clear polystyrene walls with an opaque bottom, includes on-plate spotting guides, box of 5 plates 1 * 5 items
Catalog # MSPPM768-BIO-48-5
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together