ContactUsLinkComponent
T-type thermocouples with Hastelloy wetted parts and complete with PFA holder (compression fitting) to fit DART DM7-Lee Photochem re actor. Part number TP-TCh-1/8E 1 * 1 items
  MSPPTP-TCH-1/8E
T-type thermocouples with Hastelloy wetted parts and complete with PFA holder (compression fitting) to fit DART DM7-Lee Photochem re actor. Part number TP-TCh-1/8E 1 * 1 items
  MSPPTP-TCH-1/8E
Frequently Bought Together