Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
1-Nonanol 99%

1-Nonanol 99%

Bottle, 500 g
Catalog # APOSOR70191-500G
Supplier:   Apollo Scientific
1-Nonanol 99%
Catalog # APOSOR70191-500G
Supplier:  Apollo Scientific
CAS Number:  143-08-8
1-Nonanol 99%

Bottle, 500 g

CAS Number:  143-08-8
Price
£53.80
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Nonan-1-ol, n-Nonanol, 1-Nonyl alcohol, Nonyl alcohol, 1-Hydroxy nonane, n-Nonyl alcohol, Octyl carbinol
  • CAS Number:143-08-8
  • Formula:H₃C(CH₂)₇CH₂OH
  • MDL Number:MFCD00002990
  • Molecular Weight:144.26 g/mol
  • Boiling Pt:214 °C
  • Melting Pt:–5 °C
  • Density:0,828 g/cm³ (25 °C)
  • Flash Pt:98 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications