Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

3,5-Dimethylcyclohexanone 95%

Bottle, 1 g
Catalog # APOSOR938239-1G
Supplier:   Apollo Scientific
3,5-Dimethylcyclohexanone 95%
Catalog # APOSOR938239-1G
Supplier:  Apollo Scientific
CAS Number:  2320-30-1
3,5-Dimethylcyclohexanone 95%

Bottle, 1 g

CAS Number:  2320-30-1
Price
£22.30
Log in for availability
1 * 1 g
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:2320-30-1
  • Formula:C₈H₁₄O
  • MDL Number:MFCD00019467
  • Molecular Weight:126.1962 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.