3,4'-Dihydroxypropiophenone 95%
Bottle, 250 mg
Catalog # APOSOR916419-250MG
Supplier: Apollo Scientific
3,4'-Dihydroxypropiophenone 95%
Catalog # APOSOR916419-250MG
Supplier: Apollo Scientific
CAS Number: 53170-93-7
Jump to:
Product Details & Documents
- CAS Number:53170-93-7
- Formula:C₉H₁₀O₃
- MDL Number:MFCD24688528
- Molecular Weight:166.174 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



