Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
 

2-Hydroxy-4-methylbenzonitrile 98%

Bottle, 5 g
Catalog # APOSOR903861-5G
Supplier:   Apollo Scientific
2-Hydroxy-4-methylbenzonitrile 98%
Catalog # APOSOR903861-5G
Supplier:  Apollo Scientific
CAS Number:  18495-14-2
2-Hydroxy-4-methylbenzonitrile 98%

Bottle, 5 g

CAS Number:  18495-14-2
Price
£62.90
Log in for availability
1 * 5 g
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:18495-14-2
  • Formula:C₈H₇NO
  • MDL Number:MFCD02261941
  • Molecular Weight:133.1500 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.