ContactUsLinkComponent
3’,5’-switch MOE A phosphoramidite, Rev 2’-O-MOE-A(Bz)-5’-amidite N6-Benzoyl-3’-O-(4,4’- dimethoxytrityl)-2’-O-(2-methoxyethyl)aden osine-5’-CED phosphoramidite 1 * 2 g
Catalog # MSPPGL103850-2G
3’,5’-switch MOE A phosphoramidite, Rev 2’-O-MOE-A(Bz)-5’-amidite N6-Benzoyl-3’-O-(4,4’- dimethoxytrityl)-2’-O-(2-methoxyethyl)aden osine-5’-CED phosphoramidite 1 * 2 g
Catalog # MSPPGL103850-2G
Frequently Bought Together