ContactUsLinkComponent
3’-O-MOE-G(iBu)-2’-phosphoramidite 5’-O-DMTr-N2-isobutyroyl- 3’-O-(2-methoxyethyl)guanosine-2’-CEDphosphoramidite 1 * 2 g
Catalog # MSPPGL103288-2G
3’-O-MOE-G(iBu)-2’-phosphoramidite 5’-O-DMTr-N2-isobutyroyl- 3’-O-(2-methoxyethyl)guanosine-2’-CEDphosphoramidite 1 * 2 g
Catalog # MSPPGL103288-2G
Frequently Bought Together