Parteck MXP (Polyvinyl alcohol) EMPROVE ESSENTIAL Ph Eur,JPE,USP 1 * 1 kg
Catalog # 1.41464.1000
Supplier: MERCK PRODUCTION CHEMICALS
Parteck MXP (Polyvinyl alcohol) EMPROVE ESSENTIAL Ph Eur,JPE,USP 1 * 1 kg
Catalog # 1.41464.1000
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals