Sodium dihydrogen phosphate monohydrate EMPROVE EXPERT BP,USP 1 * 12 kg
1.37093.9012
Sodium dihydrogen phosphate monohydrate EMPROVE EXPERT BP,USP 1 * 12 kg
1.37093.9012
Frequently Bought Together
Cobalt, powder, Sigma-Aldrich®
: Merck
Sodium sulfate anhydrous EMPROVE EXPERT Ph Eur,BP,ChP,USP 1 * 1 kg
: MERCK PRODUCTION CHEMICALS
Disodium hydrogen phosphate dodecahydrate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
: MERCK PRODUCTION CHEMICALS
Pesticide standard, Aldicarb, reference material, Honeywell Fluka™
: Honeywell Chemicals
Sodium Silicate neutral solution technical grade 1 * 1.000 mL
: PanReac AppliChem
Nickel (II) oxide 99.99+%
: Apollo Scientific
Thymolphthalein ACS
: Thermo Fisher Scientific