Thermometer, solid stem, similar to ASTM 132 1 290,00 53,50 134,85C, white backed, 148,6+151,4:0,05°C, /WCcapillary tube specially coated inside, withnon-wetting blue special liquid, total immersion,305x6-8mm, suitable for government calibration, dur 1 * 1 items
Catalog # AMARA301900-CB/WC
Supplier: Amarell
Frequently Bought Together