ContactUsLinkComponent
Steel wire mesh shelf unit, chrome plated, max. shelf load 100 kg, WxD 1220 x 460 mm, standard shelf unit, height 1880 mm, 5 shelves 1 * 1 items
Catalog # KAKA530965-10
Steel wire mesh shelf unit, chrome plated, max. shelf load 100 kg, WxD 1220 x 460 mm, standard shelf unit, height 1880 mm, 5 shelves 1 * 1 items
Catalog # KAKA530965-10
Frequently Bought Together