GLB1 ELISA Kit (Human)is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for GLB1 has been pre-coated onto a 96-well plate (12 x 8 Well Strips) and blocked 1 * 1 KIT
Catalog # AVIVOKCD06127
Supplier: Aviva Systems Biology