Body:316SS, MWP:1960psig, O-ring:FKM, Stem Tip:PCTFE, Conn.:1/4in. (M)NPT x 1/4in. (F)NPT, Oriffice:5.6mm, Cv:0.53, Anodized Al Hand le, With Rupture Disk:1900psig 1 * 1 items
Catalog # MSPPNDSSNS4FNS48SA
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together