X960 filtred finntip flex 100ul, sterile,Filter sterile, free from DNA, DNase and RNasein vacuum sealed sterilised tip racks polypro pylene tip, polyethylene matrix filter 10 x 96 racked 5L to 100L 1 * 960 items
Catalog # MSPP11853430
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together