Empore 4215SD: C18 bonded silica, 50um particle size. 7mm/3ml 1 * 150 items
MSPP98-0604-0198-5
Empore 4215SD: C18 bonded silica, 50um particle size. 7mm/3ml 1 * 150 items
MSPP98-0604-0198-5
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection