ContactUsLinkComponent
THIS CASSETTE IS INCLUDED IN THE NEXION 5000 ICP-MS CONFIGURATION'S SAMPLE INTRODUCTION KIT (N8160020). 1 * 1 items
Catalog # PERCN8161009
Supplier:  PerkinElmer
THIS CASSETTE IS INCLUDED IN THE NEXION 5000 ICP-MS CONFIGURATION'S SAMPLE INTRODUCTION KIT (N8160020). 1 * 1 items
Catalog # PERCN8161009
Supplier:  PerkinElmer