ContactUsLinkComponent
CORROSION-RESISTANT PROBES ARE SUITABLE FOR INORGANIC ACIDS. LENGTH: 10 IN (25.4 CM) PROBE MATERIAL: FEP-COVERED CARBON FIBER) CAPIL LARY MATERIAL: PFA) INCLUDES: PROBE WITH ATTACHED SAMPLE TUBING) THE CONTENTS OF THIS KIT ARE DESCRIBED AS FOLLOWS: 1 1 * 1 items
Catalog # PERCN0830022
Supplier:  PerkinElmer
CORROSION-RESISTANT PROBES ARE SUITABLE FOR INORGANIC ACIDS. LENGTH: 10 IN (25.4 CM) PROBE MATERIAL: FEP-COVERED CARBON FIBER) CAPIL LARY MATERIAL: PFA) INCLUDES: PROBE WITH ATTACHED SAMPLE TUBING) THE CONTENTS OF THIS KIT ARE DESCRIBED AS FOLLOWS: 1 1 * 1 items
Catalog # PERCN0830022
Supplier:  PerkinElmer
Frequently Bought Together