THE RHODAMINE 100 STANDARD IS A MIXTURE OF POLYOL (>90%) AND RHODAMINE B (<10%). IT IS PACKAGED IN A CUVETTE WITH A DIAGONAL CUTOFF AND FLAME-SEALED. ISO-CERTIFIED STANDARD. 1 * 1 items
Catalog # PERCN4202027
Supplier: PerkinElmer