ContactUsLinkComponent
Masterflex L/S® Digital Drive with Open-Head Sensor and Easy-Load® II Pump Head for High-Performance Tubing, 600 rpm 115/230 VAC 1 * 1 items
Catalog # MSPP77921-79
Masterflex L/S® Digital Drive with Open-Head Sensor and Easy-Load® II Pump Head for High-Performance Tubing, 600 rpm 115/230 VAC 1 * 1 items
Catalog # MSPP77921-79
Frequently Bought Together