ContactUsLinkComponent
Histone H4 (29- 103) [I29C] CT KPAIRRLARR GGVKRISGLI YEETRGVLKV FLENVIRDAV TYTEHAKRKT VTAMDVVYAL KRQGRTLYGF GG TFA salt 90% area purity 1 * 10 mg
Catalog # MSPPH4-I29C-10MG
Histone H4 (29- 103) [I29C] CT KPAIRRLARR GGVKRISGLI YEETRGVLKV FLENVIRDAV TYTEHAKRKT VTAMDVVYAL KRQGRTLYGF GG TFA salt 90% area purity 1 * 10 mg
Catalog # MSPPH4-I29C-10MG
Frequently Bought Together