(1-31)H3K4me1 M ARTK(Me)Q TARKS TGGKA PRKQL ATKAA RKSAP A-thioester TFA salt 95% area purity 1 * 20 mg
Catalog # MSPPH3K4ME1-20MG
Supplier: MARKET SOURCE PART PROCESS
(1-31)H3K4me1 M ARTK(Me)Q TARKS TGGKA PRKQL ATKAA RKSAP A-thioester TFA salt 95% area purity 1 * 20 mg
Catalog # MSPPH3K4ME1-20MG
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection