ContactUsLinkComponent
Microscope cover glasses, DIN ISO 8255 compliant, 42 mm Ø, borosilicate glass D 263 M, thickness No. 1.5H (0.17±0.005 mm), CE marked 1 * 1.000 items
Catalog # MARI0117760
Supplier:  MARIENFELD
Microscope cover glasses, DIN ISO 8255 compliant, 42 mm Ø, borosilicate glass D 263 M, thickness No. 1.5H (0.17±0.005 mm), CE marked 1 * 1.000 items
Catalog # MARI0117760
Supplier:  MARIENFELD
Frequently Bought Together