ContactUsLinkComponent
COVID-19 nucleoprotein ELISA Kit allows for the in vitro quantitative determination of COVID-19 nucleoprotein concentrations in plasma, tissue homogenates and other biological fluids from various mammalian species. Range: 0.0625-4 ng/mL. Sensitivity: 1 * 96 Tests
Catalog # 08440300.
Supplier:  MP Biomedicals
COVID-19 nucleoprotein ELISA Kit allows for the in vitro quantitative determination of COVID-19 nucleoprotein concentrations in plasma, tissue homogenates and other biological fluids from various mammalian species. Range: 0.0625-4 ng/mL. Sensitivity: 1 * 96 Tests
Catalog # 08440300.
Supplier:  MP Biomedicals
Frequently Bought Together