CHROMABOND gasket for lid SPE of vacuum manifold with 24 positions pack of 2 suitable for lids of REF 730151N 1 * 2 items
Catalog # MANA730178N
Supplier: MACHEREY-NAGEL
CHROMABOND gasket for lid SPE of vacuum manifold with 24 positions pack of 2 suitable for lids of REF 730151N 1 * 2 items
Catalog # MANA730178N
Supplier: MACHEREY-NAGEL
Frequently Bought Together
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
TLC plates, silica gel 60G, glass backed
Supplier: Merck
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Inserts for Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
Silica gel 60, irregular 40 - 63 µm for flash chromatography
Supplier: VWR Chemicals
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Headspace Vials ND20
Supplier: VWR Collection
Certified screw top vials and closures
Supplier: Agilent
Gold standard syringes for autosampler
Supplier: Agilent
TOC certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific