ContactUsLinkComponent
FDA conforming software AtmoCONTROL (FDA edition). Meets the requirements for the use of electronically stored data sets and electro nic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Base licence f 1 * 1 items
Catalog # MEMMFDAQ1ICO240
Supplier:  MEMMERT
FDA conforming software AtmoCONTROL (FDA edition). Meets the requirements for the use of electronically stored data sets and electro nic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Base licence f 1 * 1 items
Catalog # MEMMFDAQ1ICO240
Supplier:  MEMMERT