MICROPLATE 384 W CELLSTAR U-BOTTOM STER 1 * 32 items
Catalog # GREI787979_32
Supplier: Greiner Bio-One
MICROPLATE 384 W CELLSTAR U-BOTTOM STER 1 * 32 items
Catalog # GREI787979_32
Supplier: Greiner Bio-One
Frequently Bought Together
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: Sterilin
VWR®, Cryovials
Supplier: VWR Collection
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Transfer pipettes
Supplier: SARSTEDT
Assay microplates and blocks, 384-well, ScreenMates
Supplier: THERMO MATRIX TECHNOLOGIES
Microfuge tubes, proteomic grade
Supplier: G-Biosciences
VWR® Safe-T-Flex, Caps for Test Tubes
Supplier: VWR Collection
Storage microplates 384-well polypropylene, Corning®
Supplier: Corning
Chemical bottles, wide mouth, square
Supplier: RIXIUS
Micro packaging vials with thread, Nalgene®
Supplier: Thermo Fisher Scientific
Simplettes® for Stomacher® bags 400
Supplier: Seward (FermionX)
Accessories for cooling racks
Supplier: GLW
PCR plates, 96-well, Armadillo™
Supplier: Thermo Fisher Scientific