PARAPLAST PLUS WITH DMSO 56C 1 * 1 kg
Catalog # LEIH39602004
Supplier: LEICA HISTOLOGY
Frequently Bought Together
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
VWR® SuperFrost® Plus, Adhesion Slides
Supplier: VWR Collection
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One