ContactUsLinkComponent
Cooling Jacket. After sterilization, water is circulated in direct contact with the outside of the autoclave chamber,resulting in the rapid decrease of the internal chamber temperature. Offers a substantial time saving vs traditional cooling. Free of 1 * 1 items
Catalog # ASTLAJP152C
Supplier:  ASTELL SCIENTIFIC
Cooling Jacket. After sterilization, water is circulated in direct contact with the outside of the autoclave chamber,resulting in the rapid decrease of the internal chamber temperature. Offers a substantial time saving vs traditional cooling. Free of 1 * 1 items
Catalog # ASTLAJP152C
Supplier:  ASTELL SCIENTIFIC
Frequently Bought Together