ContactUsLinkComponent
Precision thermometer, similar DIN, -10/0+200:0,5C, enclosed scale, cap. prismatic, colourless, total immersion, red special liquid state calibrated, with DKD certificate (DAkkS) with 4 test points 1 * 1 items
Catalog # AMARL25928-FL/DKD4
Supplier:  Amarell
Precision thermometer, similar DIN, -10/0+200:0,5C, enclosed scale, cap. prismatic, colourless, total immersion, red special liquid state calibrated, with DKD certificate (DAkkS) with 4 test points 1 * 1 items
Catalog # AMARL25928-FL/DKD4
Supplier:  Amarell
Frequently Bought Together