Heated jacket. Required when sterilizing porous materials. Option required for ‘difficult’ waste loads (i.e. wrapped instruments/garments/clothing). Provides improved temperature distribution within the load. Best used with the advanced vacuum option 1 * 1 items
Catalog # ASTLAJP152
Supplier: AIR 7 SEAS TRANSPORT LOGISTICS, INC
Frequently Bought Together