ContactUsLinkComponent
24kW Integral Steam Generator (33.14 litre pressure vessel). Replaces the standard heaters in chamber. This option helps improve cycle times, and includes automatic water filling as standard (Mains supply required). Suitable for front loading models 1 * 1 items
Catalog # ASTL24KWSG
Supplier:  ASTELL SCIENTIFIC
24kW Integral Steam Generator (33.14 litre pressure vessel). Replaces the standard heaters in chamber. This option helps improve cycle times, and includes automatic water filling as standard (Mains supply required). Suitable for front loading models 1 * 1 items
Catalog # ASTL24KWSG
Supplier:  ASTELL SCIENTIFIC
Frequently Bought Together