ContactUsLinkComponent
720163.46 - EC HPLC column EC 100/4.6 NUCLEOSIL 120-5 C18 Length: 100 mm, ID: 4.6 mm pack of 1 1 * 1 items
Catalog # MANA720163.46
Supplier:  MACHEREY-NAGEL
720163.46 - EC HPLC column EC 100/4.6 NUCLEOSIL 120-5 C18 Length: 100 mm, ID: 4.6 mm pack of 1 1 * 1 items
Catalog # MANA720163.46
Supplier:  MACHEREY-NAGEL
Frequently Bought Together