Controls the reduction of chamber pressure during the cooling phase, preventing the ‘boiling over’ of bottled fluids. Requires 200lt/min comp. air (or large compressor option). NB: Load Sensed Timing is also recommended. For toploading models 95 litr 1 * 1 items
Catalog # ASTLAVC004E
Supplier: AIR 7 SEAS TRANSPORT LOGISTICS, INC
Frequently Bought Together