1920x1080 resolution 1.8GhZ quad-core processor Android-based WiFi & Ethernet VESA 100 mount pattern 0.3-1m temperature check d istance +/- 0.5C accuracy 1 * 1 items
Catalog # MSPPTTS-15
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together