ContactUsLinkComponent
Volumetric pipettes, 80,0 ml, class AS, with one mark,conformity certified, waiting time 5 s., blue DIFFICO 1 * 1 items
Catalog # WITG3.535.800.BL
Supplier:  witeg Labortechnik
Volumetric pipettes, 80,0 ml, class AS, with one mark,conformity certified, waiting time 5 s., blue DIFFICO 1 * 1 items
Catalog # WITG3.535.800.BL
Supplier:  witeg Labortechnik
Frequently Bought Together