ContactUsLinkComponent
MAXIMUM CAPACITY 220 G READABILITY 0.01 MG REPEATABILITY (TEST WEIGHT )0.015 MG (10 G) MINIMUM WEIGHT (USP, 0.1%, TYPICAL )14 MG ADJUSTMENT INTERNAL / FACT WEIGHING PAN DIMENSIONS (WXD)78 MM X 73 MM LEGAL FOR TRADE NO SETTLING TIME 2S REPEATABILITY ( 1 * 1 items
Catalog # METB30355411
Supplier:  Mettler - Toledo
MAXIMUM CAPACITY 220 G READABILITY 0.01 MG REPEATABILITY (TEST WEIGHT )0.015 MG (10 G) MINIMUM WEIGHT (USP, 0.1%, TYPICAL )14 MG ADJUSTMENT INTERNAL / FACT WEIGHING PAN DIMENSIONS (WXD)78 MM X 73 MM LEGAL FOR TRADE NO SETTLING TIME 2S REPEATABILITY ( 1 * 1 items
Catalog # METB30355411
Supplier:  Mettler - Toledo
Frequently Bought Together