Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
Collagenase, Reference Standards

Collagenase, Reference Standards

Collagenase I, USP reference standard, 2 x 0.5 mL, Vial, 1 mL
Catalog # 1148089.
Supplier:   USP
Collagenase, Reference Standards
Catalog # 1148089.
Supplier:  USP
CAS Number:  
Collagenase, Reference Standards

Collagenase I, USP reference standard, 2 x 0.5 mL, Vial, 1 mL

CAS Number:  
Price
£396.00
Log in for availability
1 * 1 mL
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mL
  • Standards:
    USP reference standard
  • Description:
    Collagenase I
  • Pk Info:
    2 x 0.5 mL
  • Cat. No.:
    1148089.
  • Description:
    Collagenase I
  • CAS No.:
    9001-12-1

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.