ContactUsLinkComponent
RF1270 LG U 1031415N5N, FREEZER, white exterior, refrigerant R290, SOLID doors, castors 360 swivel, e-sensor, 5 plastic coated wire shelves, 5 plastic coated wire shelves Including 5 years warranty on cabinets 1 * 1 items
Catalog # GRAM882700254
Supplier:  GRAM
RF1270 LG U 1031415N5N, FREEZER, white exterior, refrigerant R290, SOLID doors, castors 360 swivel, e-sensor, 5 plastic coated wire shelves, 5 plastic coated wire shelves Including 5 years warranty on cabinets 1 * 1 items
Catalog # GRAM882700254
Supplier:  GRAM
Frequently Bought Together