ContactUsLinkComponent
Chromatographic-columns, 430 ml, 30 x 600 mm, fused-in sintered glass- disc P0, w. ST-socket 29/32, ST-PTFE-stopcock, bore: 2,5 mm 1 * 1 items
Catalog # WITG2.133.013
Supplier:  witeg Labortechnik
Chromatographic-columns, 430 ml, 30 x 600 mm, fused-in sintered glass- disc P0, w. ST-socket 29/32, ST-PTFE-stopcock, bore: 2,5 mm 1 * 1 items
Catalog # WITG2.133.013
Supplier:  witeg Labortechnik
Frequently Bought Together