Polysorbate 80, GPR RECTAPUR®
Plastic bottle, 2,5 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Merck Index:13,07664
- Boiling Pt:>100 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:1,062 g/cm³ (20 °C)
- Flash Pt:149 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:500-019-9
Documents
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000)
Packaging Information
P20_VWRC28830.260_SPEC_EN.pdf P20_VWRC28830.291_SPEC_EN.pdf P20_VWRC28830.320_SPEC_EN.pdf P20_VWRC28830.460_SPEC_EN.pdf spec_VWRC28830.260
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Identification:Passes test
- Density (20/4):1.070 - 1.090
- Hydroxyl value:65 - 80
- Saponification value:45 - 55
- Acid value:Max 2 mg KOH/g
- Water content:Max 3%
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



