10 µg-The Thermo Scientific pEF1α Tluc16 Vector encodes the smallest known luciferase, TurboLuc16 (Tluc16) luciferase, as a reporter under a modified version of the Elongated Factor 1α (EF1α) promoter for constitutive Tluc16 luciferase expression in 1 * 10 µG
Catalog # PIER88241
Supplier: Thermo Fisher Scientific