ContactUsLinkComponent
A prodrug of benazeprilat inhibits enzymatic activity of partially purified ACE isolated from rabbit lung (IC50 = 2 nM) 1 * 50 mg
Catalog # CAYM23804-50
Supplier:  Cayman Chemical
A prodrug of benazeprilat inhibits enzymatic activity of partially purified ACE isolated from rabbit lung (IC50 = 2 nM) 1 * 50 mg
Catalog # CAYM23804-50
Supplier:  Cayman Chemical
Frequently Bought Together