ContactUsLinkComponent
Troemner OIML Precision Weights Wt Set(28), 5 kg-1 mg, OIML E1, NVLAP 1 * 1 items
Catalog # OHAU30390912
Supplier:  OHAUS
Troemner OIML Precision Weights Wt Set(28), 5 kg-1 mg, OIML E1, NVLAP 1 * 1 items
Catalog # OHAU30390912
Supplier:  OHAUS
Frequently Bought Together