VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 100075 MPA.S (25.00C), RT100000 1 * 600 mL
Catalog # PARGVIS-RT100K-600
Supplier: PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 100075 MPA.S (25.00C), RT100000 1 * 600 mL
Catalog # PARGVIS-RT100K-600
Supplier: PARAGON SCIENTIFIC
Frequently Bought Together
Phosphorus CRM, standard solutions, Supelco®
Supplier: Merck
Acetic acid 12% Reag. Ph. Eur. 1000402
Supplier: VWR Chemicals
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur. 1091201
Supplier: VWR Chemicals
Humidity standards
Supplier: ROTRONIC INSTRUMENTS
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, TOC standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
pH buffer solutions
Supplier: Mettler - Toledo
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Organic reference standard, Trolamine, Ph.Eur., RS
Supplier: EDQM
VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography
Supplier: VWR Chemicals