ContactUsLinkComponent
Dust cover for Sartorius Entris® analytical and milligram balances with an analytical draft shield with a height of 240 mm. For Sartorius lab balances of the product family Entris® with a readability of 0.1 mg and 1 mg. 1 * 1 items
Catalog # SARB6960BC01
Supplier:  Sartorius Balances
Dust cover for Sartorius Entris® analytical and milligram balances with an analytical draft shield with a height of 240 mm. For Sartorius lab balances of the product family Entris® with a readability of 0.1 mg and 1 mg. 1 * 1 items
Catalog # SARB6960BC01
Supplier:  Sartorius Balances
Frequently Bought Together