ContactUsLinkComponent
Potassium Permanganate 0.2M (1N). Duree de conservation : 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 kg
Catalog # CMPLZ14641079.1L
Potassium Permanganate 0.2M (1N). Duree de conservation : 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 1 kg
Catalog # CMPLZ14641079.1L
Frequently Bought Together