ContactUsLinkComponent
Surface protection low adhesion cleanroom tape, model 1310, PVC backing, acrylic adhesive, transparent blue, width 1/2 (12,7mm), length 55 yards (50,3m), Ultra Clean, double-bagged 1 * 12 Roll
Catalog # UTAP1310TB050-P5S
Supplier:  ULTRATAPE
Surface protection low adhesion cleanroom tape, model 1310, PVC backing, acrylic adhesive, transparent blue, width 1/2 (12,7mm), length 55 yards (50,3m), Ultra Clean, double-bagged 1 * 12 Roll
Catalog # UTAP1310TB050-P5S
Supplier:  ULTRATAPE