ContactUsLinkComponent
Cubis MCA balance with large high end 7” color TFT touch display in 16:9 format with new user interface. Actual scale division (d) 1 0 mg, nominal maximum load (Max) 1200g. Standard version non-verified, all units. Flat, stainless steel weight pan wi 1 * 1 items
Catalog # SARBMCA1202S2S00-0
Supplier:  Sartorius Balances
Cubis MCA balance with large high end 7” color TFT touch display in 16:9 format with new user interface. Actual scale division (d) 1 0 mg, nominal maximum load (Max) 1200g. Standard version non-verified, all units. Flat, stainless steel weight pan wi 1 * 1 items
Catalog # SARBMCA1202S2S00-0
Supplier:  Sartorius Balances
Frequently Bought Together